Propylbenzene ≥99.0%
Supplier: TCI America
|
Danger
|
P0523-500ML
P0523-25ML
P0523-100ML
TCP0523-500MLEA
357
USD
TCP0523-500ML
TCP0523-25ML
TCP0523-100ML
Propylbenzene ≥99.0%
Propylbenzene
CAS Number: 103-65-1
MDL Number: MFCD00009377
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 159
Melting point (°C): -99
Flash Point (°C): 30
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: 1-Phenylpropane
Formula:
C₉H₁₂ MW: 120.19 g/mol Boiling Pt: 159 °C Melting Pt: –99 °C Density: 0.86 (20 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009377 CAS Number: 103-65-1 EINECS: 203-132-9 UN: 2364 ADR: 3,III Merck Index: 12,08029 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...