Propylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
1-Propylamine, 1-Aminopropane
P0520-500ML
P0520-25ML
TCP0520-500MLEA
42
USD
TCP0520-500ML
TCP0520-25ML
Propylamine ≥98.0% (by GC, titration analysis)
Propylamine
CAS Number: 107-10-8
MDL Number: MFCD00008205
Molecular Formula: C3H9N
Molecular Weight: 59.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 48
Melting point (°C): -83
Flash Point (°C): -30
Specific Gravity (20/20): 0.72
Other Product Names / Synonyms: 1-Aminopropane
Formula:
H₃CCH₂CH₂NH₂ MW: 59.11 g/mol Boiling Pt: 48 °C Melting Pt: –83 °C Density: 0.72 (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008205 CAS Number: 107-10-8 EINECS: 203-462-3 UN: 1277 ADR: 3,II Merck Index: 12,08028 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...