Propiononitrile ≥99.0%
Supplier: TCI America
Synonyms:
Propionitrile
P0514-500ML
P0514-25ML
P0514-100ML
TCP0514-500MLEA
69.39
USD
TCP0514-500ML
TCP0514-25ML
TCP0514-100ML
Propiononitrile ≥99.0%
Propiononitrile
CAS Number: 107-12-0
MDL Number: MFCD00001948
Molecular Formula: C3H5N
Molecular Weight: 55.08
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 13
Specific Gravity (20/20): 0.78
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Ethyl Cyanide
Formula:
C₃H₅N MW: 55.08 g/mol Boiling Pt: 96 °C Melting Pt: –93 °C Density: 0.78 (20 °C) Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001948 CAS Number: 107-12-0 EINECS: 203-464-4 UN: 2404 ADR: 3,II Merck Index: 12,08012 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...