Propyl propionate ≥98.0%

Supplier: TCI America
Danger

P0511-25ML P0511-500ML
TCP0511-025MLEA 16.79 USD
TCP0511-025ML TCP0511-500ML
Propyl propionate ≥98.0%
Propyl propionate

CAS Number: 106-36-5
MDL Number: MFCD00009373
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 27
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Propionic Acid Propyl Ester

Formula: C₆H₁₂O₂
Boiling Pt: 122 °C
Density: 0.88 (20 °C)
Flash Pt: 27 °C
MDL Number: MFCD00009373
CAS Number: 106-36-5
EINECS: 203-389-7
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9ce4cfa8ca10e74e4fb1ff9314ecdd666ab10b5&vid=b9ce4cfa8ca10e74e4fb1ff9314ecdd666ab10b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR