Methyl propionate ≥99.0%

Supplier: TCI America
Danger

Synonyms: Propanoic acid methyl ester

P0508-500ML P0508-25ML
TCP0508-500MLEA 80.59 USD
TCP0508-500ML TCP0508-25ML
Methyl propionate ≥99.0%
Methyl propionate

CAS Number: 554-12-1
MDL Number: MFCD00009306
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Melting point (°C): -88
Flash Point (°C): -4
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Propionic Acid Methyl Ester

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80 °C
Melting Pt: –88 °C
Density: 0.92 (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009306
CAS Number: 554-12-1
EINECS: 209-060-4
UN: 1248
ADR: 3,II
Merck Index: 12,06191
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee64c0d09794e86969c8bf59d36644293bf59f88&vid=ee64c0d09794e86969c8bf59d36644293bf59f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR