trans-Anethole ≥98.0%
Supplier: TCI America
CAS Number: 4180-23-8
MDL Number: MFCD00009284
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 92
Freezing point (°C): 20
Specific Gravity (20/20): 0.99
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-p-Methoxypropenylbenzene, trans-p-Propenylanisole
Formula:
C₁₀H₁₂O Boiling Pt: 235 °C Melting Pt: 21…23 °C Density: 0.99 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00009284 CAS Number: 4180-23-8 EINECS: 224-052-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...