1-Propanethiol ≥98.0%

Supplier: TCI America
Danger

Synonyms: n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane

P0488-500ML P0488-25ML
TCP0488-500MLEA 89 USD
TCP0488-500ML TCP0488-25ML
1-Propanethiol ≥98.0%
1-Propanethiol

CAS Number: 107-03-9
MDL Number: MFCD00004900
Molecular Formula: C3H8S
Molecular Weight: 76.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Melting point (°C): -113
Flash Point (°C): -20
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: Propyl Mercaptan

Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 67 °C
Melting Pt: –113 °C
Density: 0.84 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004900
CAS Number: 107-03-9
EINECS: 203-455-5
UN: 2402
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87fed1ad239cf7e56cdd2d7766e0d69befab2d09&vid=deb969c63bcb5e117cc19cbb8d50a41ae9f6cf21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR