Piperazine hexahydrate ≥97.0%

Supplier: TCI America
Danger

Synonyms: Piperazine hydrate

P0447-25G P0447-500G
TCP0447-25GEA 21.37 USD
TCP0447-25G TCP0447-500G
Piperazine hexahydrate ≥97.0%
Piperazine hexahydrate

CAS Number: 142-63-2
MDL Number: MFCD00149389
Molecular Formula: C4H10N2
Molecular Weight: 86.14
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 87


Other Product Names / Synonyms: Diethylenediamine

Formula: C₄H₁₀N₂·6H₂O
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: 44 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00149389
CAS Number: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 12,07617
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f04312946206d32114ba274e8cdc67e71e26e9&vid=e2f04312946206d32114ba274e8cdc67e71e26e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR