Pyridine-2-carbaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde
P0425-25G
P0425-500G
TCP0425-025GEA
71.1
USD
TCP0425-025G
TCP0425-500G
Pyridine-2-carbaldehyde ≥98.0%
Pyridine-2-carbaldehyde
CAS Number: 1121-60-4
MDL Number: MFCD00006290
Molecular Formula: C6H5NO
Molecular Weight: 107.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Melting point (°C): -21
Flash Point (°C): 77
Specific Gravity (20/20): 1.12
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: Picolinaldehyde
Formula:
C₆H₅NO Boiling Pt: 181 °C Melting Pt: -21 °C Density: 1.12 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00006290 CAS Number: 1121-60-4 EINECS: 214-333-6 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...