4-Picoline ≥98.0%

Supplier: TCI America

Synonyms: 4-Methylpyridine, γ-Picoline

P0417-500ML P0417-25ML
TCP0417-500MLEA 36.61 USD
TCP0417-500ML TCP0417-25ML
4-Picoline ≥98.0%
4-Picoline

CAS Number: 108-89-4
MDL Number: MFCD00006440
Molecular Formula: C6H7N
Molecular Weight: 93.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Melting point (°C): 4
Flash Point (°C): 37
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 4-Picoline

Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 144.9
Melting Pt: 3
Density: 0.96 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00006440
CAS Number: 108-89-4
EINECS: 203-626-4
UN: 2313
ADR: 3 III
Merck Index: 12,07556
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=402f294dfaadae030425c8ae49733cefc27a33f4&vid=402f294dfaadae030425c8ae49733cefc27a33f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR