2-Picoline ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2-Methylpyridine, α-Picoline
P0415-25ML
P0415-500ML
TCP0415-25MLEA
25
USD
TCP0415-25ML
TCP0415-500ML
2-Picoline ≥98.0%
2-Picoline
CAS Number: 109-06-8
MDL Number: MFCD00006332
Molecular Formula: C6H7N
Molecular Weight: 93.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 128
Melting point (°C): -70
Flash Point (°C): 25
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 2-Picoline
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 129.5 Melting Pt: -70 Density: 0.95 (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006332 CAS Number: 109-06-8 EINECS: 203-643-7 UN: 2313 ADR: 3 III Merck Index: 12,07554 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...