Phthalonitrile ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene

P0404-25G P0404-100G
TCP0404-25GEA 17 USD
TCP0404-25G TCP0404-100G
Phthalonitrile ≥99.0%
Phthalonitrile

CAS Number: 91-15-6
MDL Number: MFCD00001771
Molecular Formula: C8H4N2
Molecular Weight: 128.13
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 140
Flash Point (°C): 162


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyanobenzene

Formula: C₈H₄N₂
MW: 128.13 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 140 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00001771
CAS Number: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbc5855a688853d5d932798b27c8a8fe49f318ff&vid=dbc5855a688853d5d932798b27c8a8fe49f318ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR