1,3-Propanesultone ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Oxathiolane 2,2-dioxide

P0324-500G P0324-25G
TCP0324-500GEA 370.11 USD
TCP0324-500G TCP0324-25G
1,3-Propanesultone ≥99.0%
1,3-Propanesultone

CAS Number: 1120-71-4
MDL Number: MFCD00005355
Molecular Formula: C3H6O3S
Molecular Weight: 122.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 180
Flash Point (°C): 110
Freezing point (°C): 32


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,2-Oxathiolane 2,2-Dioxide

Formula: C₃H₆O₃S
MW: 122.14 g/mol
Boiling Pt: 180 °C
Melting Pt: 31 °C
Density: 1.3996 g/cm³ (35 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3018a5589ca511345fb665f0a66de8a056085ff&vid=b3018a5589ca511345fb665f0a66de8a056085ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR