3,5-Dimethoxyphenol ≥98.0% (by GC)

Supplier: TCI America
Warning

P0320-25G P0320-100G
TCP0320-025GEA 150 USD
TCP0320-025G TCP0320-100G
3,5-Dimethoxyphenol ≥98.0% (by GC)
3,5-Dimethoxyphenol

CAS Number: 500-99-2
MDL Number: MFCD00008388
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 45
Flash Point (°C): 78


Other Product Names / Synonyms: Phloroglucinol Dimethyl Ether

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 160 °C
Melting Pt: 45 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008388
CAS Number: 500-99-2
EINECS: 207-917-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dee5e9d5380eba5ac5eca5ac9cdc83d23eab5fb&vid=3dee5e9d5380eba5ac5eca5ac9cdc83d23eab5fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR