1-Pentene ≥98.0%

Supplier: TCI America

Synonyms: alpha-n-Amylene, Propylethylene

P0316-500ML P0316-25ML
TCP0316-500MLEA 375 USD
TCP0316-500ML TCP0316-25ML
1-Pentene ≥98.0%
1-Pentene

CAS Number: 109-67-1
MDL Number: MFCD00003567
Molecular Formula: C5H10
Molecular Weight: 70.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 30
Flash Point (°C): -51
Specific Gravity (20/20): 0.64
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Amylene

Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 30 °C
Melting Pt: –165 °C
Density: 0.64 (20 °C)
Flash Pt: –51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003567
CAS Number: 109-67-1
EINECS: 203-694-5
UN: 1108
ADR: 3,I
Merck Index: 12,07262
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2f594b8ea703e3e1e899b9f4523a4bcff16ae4d&vid=18bc0ccfa7de572862e0d92a7f477b6fc8dd0339&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR