Bis(2-ethylhexyl)phthalate ≥98.0%

Supplier: TCI America
Danger

P0297-500G P0297-25G
TCP0297-500GEA 27.47 USD
TCP0297-500G TCP0297-25G
Bis(2-ethylhexyl)phthalate ≥98.0%
Bis(2-ethylhexyl)phthalate

CAS Number: 117-81-7
MDL Number: MFCD00009493
Molecular Formula: C24H38O4
Molecular Weight: 390.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 361
Melting point (°C): -50
Flash Point (°C): 218
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Di(2-ethylhexyl) Phthalate, Dioctyl Phthalate, Phthalic Acid Bis(2-ethylhexyl) Ester, Phthalic Acid Di(2-ethylhexyl) Ester, Phthalic Acid Dioctyl Ester, DEHP, DOP

Formula: C₂₄H₃₈O₄
Boiling Pt: 361 °C
Melting Pt: –50 °C
Density: 0.99 (20 °C)
Flash Pt: 218 °C
Storage Temperature: Ambient
MDL Number: MFCD00009493
CAS Number: 117-81-7
EINECS: 204-211-0
UN: 3082
ADR: 9,III
Merck Index: 13,02884
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2f3660c470b8c4eb1b18709e47fa57a25195b28&vid=a2f3660c470b8c4eb1b18709e47fa57a25195b28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR