Dipentyl phthalate ≥98.0%

Supplier: TCI America

Synonyms: Di-n-pentyl phthalate

P0291-25G P0291-500G P0291-100G
TCP0291-025GEA 77.58 USD
TCP0291-025G TCP0291-500G TCP0291-100G
Dipentyl phthalate ≥98.0%
Dipentyl phthalate

CAS Number: 131-18-0
MDL Number: MFCD00041934
Molecular Formula: C18H26O4
Molecular Weight: 306.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 342
Flash Point (°C): 118
Specific Gravity (20/20): 1.03


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Dipentyl Phthalate, Phthalic Acid Diamyl Ester, Phthalic Acid Dipentyl Ester

Formula: C₁₈H₂₆O₄
MW: 306.41 g/mol
Boiling Pt: 342 °C
Density: 1.03 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00041934
CAS Number: 131-18-0
EINECS: 205-017-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c69c85cc84f568f6f71c489b4bbbb1e7fe6629c&vid=7c69c85cc84f568f6f71c489b4bbbb1e7fe6629c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR