Diallyl phthalate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Phthalic acid diallyl ester

P0290-500ML P0290-25ML
TCP0290-500MLEA 43 USD
TCP0290-500ML TCP0290-25ML
Diallyl phthalate ≥98.0%
Diallyl phthalate

CAS Number: 131-17-9
MDL Number: MFCD00008646
Molecular Formula: C14H14O4
Molecular Weight: 246.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 163
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Phthalic Acid Diallyl Ester

Formula: C₁₄H₁₄O₄
MW: 246.26 g/mol
Boiling Pt: 158 °C
Melting Pt: –70 °C
Density: 1.12 (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00008646
CAS Number: 131-17-9
EINECS: 205-016-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3efd66b55dece19785e821a6f0fd0089d3ac1137&vid=3efd66b55dece19785e821a6f0fd0089d3ac1137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR