Tritolyl phosphate (mixture of isomers) ≥99.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Tricresyl phosphate, Tritolyl phosphate, Tris(methylphenyl) phosphate, TCP
CAS Number: 1330-78-5
MDL Number: MFCD02102116
Molecular Formula: C21H21O4P
Molecular Weight: 368.37
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Tritolyl Ester, Phosphoric Acid Tricresyl Ester, Tritolyl Phosphate
Formula:
C₂₁H₂₁O₄P MW: 368.37 g/mol Boiling Pt: 420 Melting Pt: -40 Density: 1.17 (20 °C) Flash Pt: 234 °C |
MDL Number:
MFCD02102116 CAS Number: 1330-78-5 EINECS: 215-548-8 UN: 2810 ADR: 6.1 II Merck Index: 12,09892 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...