Triphenyl phosphate ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Triphenylphosphate, TPP
P0272-500G
P0272-25G
TCP0272-500GEA
46.36
USD
TCP0272-500G
TCP0272-25G
Triphenyl phosphate ≥99.0%
Triphenyl phosphate
CAS Number: 115-86-6
MDL Number: MFCD00003031
Molecular Formula: C18H15O4P
Molecular Weight: 326.29
Purity/Analysis Method: >99.0% (GC)
Form: Flake
Boiling point (°C): 220
Melting point (°C): 50
Flash Point (°C): 220
Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Resin Additives, Resin Additives
Other Product Names / Synonyms: Phosphoric Acid Triphenyl Ester
Formula:
C₁₈H₁₅O₄P MW: 326.29 g/mol Boiling Pt: 412 Melting Pt: 50 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003031 CAS Number: 115-86-6 EINECS: 204-112-2 UN: 3077 ADR: 9 III Merck Index: 12,09872 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...