N,N,N-Trimethylanilinium bromide ≥98.0%
Supplier: TCI America
Synonyms:
Phenyltrimethylammonium bromide, Trimethylphenylammonium bromide
P0243-500G
P0243-25G
P0243-100G
TCP0243-500GEA
259
USD
TCP0243-500G
TCP0243-025G
TCP0243-100G
N,N,N-Trimethylanilinium bromide ≥98.0%
N,N,N-Trimethylanilinium bromide
CAS Number: 16056-11-4
MDL Number: MFCD00011788
Molecular Formula: C9H14BrN
Molecular Weight: 216.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: Phenyltrimethylammonium Bromide
Formula:
C₉H₁₄BrN Melting Pt: 214 °C |
MDL Number:
MFCD00011788 CAS Number: 16056-11-4 EINECS: 240-202-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...