1-Phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: N-Phenylthiourea, 1-Phenylthiourea

P0237-500G P0237-25G
TCP0237-500GEA 421 USD
TCP0237-500G TCP0237-25G
1-Phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-Phenyl-2-thiourea

CAS Number: 103-85-5
MDL Number: MFCD00004933
Molecular Formula: C7H8N2S
Molecular Weight: 152.22
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 153


Product Categories: Synthetic Organic Chemistry, Thioureas (Multi-component Reaction)
Other Product Names / Synonyms: Phenylthiocarbamide, PTC

Formula: C₇H₈N₂S
MW: 152.22 g/mol
Boiling Pt: 295…297 °C (1013 hPa)
Melting Pt: 153 °C
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I
Merck Index: 12,07468
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=472d4a297d0f402658d0dbf8d877a950cc566a65&vid=472d4a297d0f402658d0dbf8d877a950cc566a65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR