Diphenyl sulfide ≥98.0%
Supplier: TCI America
|
Warning
|
CAS Number: 139-66-2
MDL Number: MFCD00003064
Molecular Formula: C12H10S
Molecular Weight: 186.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 296
Melting point (°C): -40
Flash Point (°C): 126
Specific Gravity (20/20): 1.12
Product Categories: Materials Chemistry, Diphenyl Sulfides (for High-Performance Polymer Research)
Other Product Names / Synonyms: Diphenyl Sulfide
Formula:
C₁₂H₁₀S Boiling Pt: 296 °C Melting Pt: –40 °C Density: 1.12 (20 °C) Flash Pt: 126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003064 CAS Number: 139-66-2 EINECS: 205-371-4 UN: 2810 ADR: 6.1,III Merck Index: 12,07467 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...