m-Phenylenediamine dihydrochloride ≥99.0%

Supplier: TCI America
Danger

Synonyms: Benzene-1,3-diamine dihydrochloride, 1,3-Diaminobenzene dihydrochloride, 1,3-Phenylenediamine dihydrochloride, 1,3-Phenylenediammonium dichloride

P0172-500G P0172-25G
TCP0172-500GEA 348.4 USD
TCP0172-500G TCP0172-25G
m-Phenylenediamine dihydrochloride ≥99.0%
m-Phenylenediamine dihydrochloride

CAS Number: 541-69-5
MDL Number: MFCD00012975
Molecular Formula: C6H8N2
Molecular Weight: 181.06
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Other Product Names / Synonyms: 1,3-Diaminobenzene Dihydrochloride

Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 277…278 °C
Storage Temperature: Ambient
MDL Number: MFCD00012975
CAS Number: 541-69-5
EINECS: 208-790-0
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5b28376ee692e94f8e1374947b8f95ecbcfc724&vid=c5b28376ee692e94f8e1374947b8f95ecbcfc724&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR