Benzylideneacetone ≥98.0%

Supplier: TCI America

Synonyms: 4-Phenylbut-3-en-2-one, 4-Phenyl-3-buten-2-one

P0163-25G P0163-500G
TCP0163-025GEA 28.03 USD
TCP0163-025G TCP0163-500G
Benzylideneacetone ≥98.0%
Benzylideneacetone

CAS Number: 122-57-6
MDL Number: MFCD00008779
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40
Flash Point (°C): 66


Other Product Names / Synonyms: Benzylideneacetone, Methyl Styryl Ketone, 4-Phenyl-3-buten-2-one

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Melting Pt: 40 °C
Flash Pt: 66 °C
MDL Number: MFCD00008779
CAS Number: 122-57-6
EINECS: 204-555-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35d47d30b26fbaaa92b02b4cffa2d43354b422bb&vid=35d47d30b26fbaaa92b02b4cffa2d43354b422bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR