Phenylacetyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P0129-500G P0129-25G P0129-100G
TCP0129-500GEA 347 USD
TCP0129-500G TCP0129-25G TCP0129-100G
Phenylacetyl chloride ≥98.0% (by GC, titration analysis)
Phenylacetyl chloride

CAS Number: 103-80-0
MDL Number: MFCD00000729
Molecular Formula: C8H7ClO
Molecular Weight: 154.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 102
Specific Gravity (20/20): 1.17

Formula: C₈H₇ClO
Boiling Pt: 211 °C
Melting Pt: 91 °C
Density: 1.17 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00000729
CAS Number: 103-80-0
EINECS: 203-146-5
UN: 2577
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9d3296260517e0e331be30bcf71b4fbc6918a71&vid=683b46a2daab13b64171cabfd5fc762058ebf1b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR