2-Pentanone ≥99.0%

Supplier: TCI America
Danger

Synonyms: Methyl Propyl Ketone

P0060-500ML P0060-25ML
TCP0060-500MLEA 117.76 USD
TCP0060-500ML TCP0060-25ML
2-Pentanone ≥99.0%
2-Pentanone

CAS Number: 107-87-9
MDL Number: MFCD00009400
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Melting point (°C): -78
Flash Point (°C): 7
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Methyl Propyl Ketone

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 101 °C
Melting Pt: –78 °C
Density: 0.81 (20 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3,II
Merck Index: 12,06193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1da5d5132b8900f107c2e74cb679fa212d90a13&vid=aa6ce9f47506fda4d9afc57146f8f4fa5cf94913&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR