1-Pentanol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol
CAS Number: 71-41-0
MDL Number: MFCD00002977
Molecular Formula: C5H12O
Molecular Weight: 88.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Melting point (°C): -79
Flash Point (°C): 43
Specific Gravity (20/20): 0.82
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Analytical Science, Odor Deterioration Factors of Cosmetics
Other Product Names / Synonyms: Amyl Alcohol, Butylcarbinol
Formula:
H₃C(CH₂)₃CH₂OH Boiling Pt: 137 °C Melting Pt: -79 °C Density: 0.82 (20 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002977 CAS Number: 71-41-0 EINECS: 200-752-1 UN: 1105 ADR: 3,III Merck Index: 13,07195 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...