Acetylacetone ≥99.0%

Supplier: TCI America
Danger

P0052-500ML P0052-25ML
TCP0052-500MLEA 68.67 USD
TCP0052-500ML TCP0052-25ML
Acetylacetone ≥99.0%
Acetylacetone

CAS Number: 123-54-6
MDL Number: MFCD00008787
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Melting point (°C): -23
Flash Point (°C): 34
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands
Other Product Names / Synonyms: Diacetylmethane, 2,4-Pentanedione

Formula: CH₃COCH₂COCH₃
Boiling Pt: 138 °C
Melting Pt: –23 °C
Density: 0.97 (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3,III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78079845ff80daac31c096a1fed9ad46c0e5ee31&vid=c66f098bd40083889ce233eb2fa54f111948e6ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR