Octaphenylcyclotetrasiloxane ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
2,2,4,4,6,6,8,8-Octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
O0231-25G
O0231-5G
TCO0231-025GEA
62.07
USD
TCO0231-025G
TCO0231-5G
Octaphenylcyclotetrasiloxane ≥98.0% (by HPLC)
Octaphenylcyclotetrasiloxane
CAS Number: 546-56-5
MDL Number: MFCD00003268
Molecular Formula: C48H40O4Si4
Molecular Weight: 793.18
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Boiling point (°C): 332
Melting point (°C): 201
Flash Point (°C): 200
Formula:
C₄₈H₄₀O₄Si₄ MW: 793.19 g/mol Boiling Pt: 332 °C Melting Pt: 201 °C Density: 1.089 g/cm³ (20 °C) Flash Pt: 200 °C |
MDL Number:
MFCD00003268 CAS Number: 546-56-5 EINECS: 208-904-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...