4'-Octylacetophenone ≥97.0%

Supplier: TCI America

Synonyms: 1-(4-Octylphenyl)ethanone, p-Octylacetophenone

O0207-25G O0207-5G
TCO0207-025GEA 317 USD
TCO0207-025G TCO0207-5G
4'-Octylacetophenone ≥97.0%
4'-Octylacetophenone

CAS Number: 10541-56-7
MDL Number: MFCD00043682
Molecular Formula: C16H24O
Molecular Weight: 232.37
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Freezing point (°C): 23

Formula: C₁₆H₂₄O
MW: 232.37 g/mol
MDL Number: MFCD00043682
CAS Number: 10541-56-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c176efa353d2996364fc490cc9d4c457f6aa2f7&vid=6c176efa353d2996364fc490cc9d4c457f6aa2f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR