Octanophenone ≥98.0%

Supplier: TCI America

Synonyms: n-Octanophenone

O0149-25ML O0149-500ML O0149-100ML
TCO0149-025MLEA 43 USD
TCO0149-025ML TCO0149-500ML TCO0149-100ML
Octanophenone ≥98.0%
Octanophenone

CAS Number: 1674-37-9
MDL Number: MFCD00003554
Molecular Formula: C14H20O
Molecular Weight: 204.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 290
Flash Point (°C): 110
Freezing point (°C): 22


Other Product Names / Synonyms: Heptyl Phenyl Ketone

Formula: C₆H₅C(O)(CH₂)₆CH₃
MW: 204.31 g/mol
Boiling Pt: 290 °C
Melting Pt: 22 °C
Density: 0.9434 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003554
CAS Number: 1674-37-9
EINECS: 216-817-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ab943b2dd985544f00861918259616b388e4c48&vid=cb62018b9d8858070f9b0a86768afc6e13f18538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR