trans-4-Octene ≥99.0%
Supplier: TCI America
Synonyms:
(E)-4-Octene
O0136-1ML
O0136-5ML
TCO0136-001MLEA
58
USD
TCO0136-001ML
TCO0136-5ML
trans-4-Octene ≥99.0%
trans-4-Octene
CAS Number: 14850-23-8
MDL Number: MFCD00009476
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 21
Specific Gravity (20/20): 0.71
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 122 °C Melting Pt: –93.8 °C Density: 0.71 (20 °C) Flash Pt: 21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009476 CAS Number: 14850-23-8 EINECS: 238-913-3 UN: 3295 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...