Trimethyl orthoacetate ≥98.0%

Supplier: TCI America

O0115-25ML O0115-500ML
TCO0115-025MLEA 31.4 USD
TCO0115-025ML TCO0115-500ML
Trimethyl orthoacetate ≥98.0%
Trimethyl orthoacetate

CAS Number: 1445-45-0
MDL Number: MFCD00008477
Molecular Formula: C5H12O3
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 16
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Orthoacetic Acid Trimethyl Ester, 1,1,1-Trimethoxyethane

Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 108 °C
Melting Pt: –58 °C
Density: 0.96 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00008477
CAS Number: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77195596c50ae3fd6535688b726a66f1455d39de&vid=77195596c50ae3fd6535688b726a66f1455d39de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR