4,4'-Diaminodiphenyl ether ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4,4'-Diaminodiphenyl ether
CAS Number: 101-80-4
MDL Number: MFCD00007863
Molecular Formula: C12H12N2O
Molecular Weight: 200.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 192
Flash Point (°C): 218
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Diaminodiphenyl Oxide, 4,4'-Oxydianiline
Formula:
C₁₂H₁₂N₂O MW: 200.24 g/mol Boiling Pt: 389 °C (1013 hPa) Melting Pt: 192 °C Flash Pt: 218 °C |
MDL Number:
MFCD00007863 CAS Number: 101-80-4 EINECS: 202-977-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...