1,2-Epoxycyclohexane ≥98.0%

Supplier: TCI America
Danger

Synonyms: Epoxycyclohexane, Cyclohexene oxide

O0071-500ML O0071-25ML O0071-100ML
TCO0071-500MLEA 82 USD
TCO0071-500ML TCO0071-25ML TCO0071-100ML
1,2-Epoxycyclohexane ≥98.0%
1,2-Epoxycyclohexane

CAS Number: 286-20-4
MDL Number: MFCD00005162
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 27
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Cyclohexene Oxide, 7-Oxabicyclo[4.1.0]heptane

Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 130 °C
Melting Pt: –27 °C
Density: 0.97 (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e40dccf40de83b71fcc76048d7e126ab99fc520&vid=9e40dccf40de83b71fcc76048d7e126ab99fc520&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR