Trimethyl orthoformate ≥98.0%

Supplier: TCI America

O0068-25ML O0068-500ML
TCO0068-025MLEA 22.39 USD
TCO0068-025ML TCO0068-500ML
Trimethyl orthoformate ≥98.0%
Trimethyl orthoformate

CAS Number: 149-73-5
MDL Number: MFCD00008483
Molecular Formula: C4H10O3
Molecular Weight: 106.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): -53
Flash Point (°C): 15
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, Formylation, Alkylation (Esterification)
Other Product Names / Synonyms: Methyl Orthoformate, Orthoformic Acid Trimethyl Ester, Trimethoxymethane

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 100 °C
Melting Pt: –53 °C
Density: 0.97 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II
Merck Index: 12,07012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77041bf6d4924558373f99b6c27efcdec399ff8b&vid=77041bf6d4924558373f99b6c27efcdec399ff8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR