Triethyl orthopropionate ≥96.0%

Supplier: TCI America
Danger

O0067-500ML O0067-25ML
TCO0067-500MLEA 173 USD
TCO0067-500ML TCO0067-025ML
Triethyl orthopropionate ≥96.0%
Triethyl orthopropionate

CAS Number: 115-80-0
MDL Number: MFCD00009226
Molecular Formula: C9H20O3
Molecular Weight: 176.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 47
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Ethyl Orthopropionate, Orthopropionic Acid Triethyl Ester, 1,1,1-Triethoxypropane

Formula: C₉H₂₀O₃
MW: 176.26 g/mol
Boiling Pt: 158 °C
Melting Pt: < 0 °C
Density: 0.89 (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00009226
CAS Number: 115-80-0
EINECS: 204-108-0
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c4841a5da826ee723e5f127c01218bf8aaafa8d&vid=5c4841a5da826ee723e5f127c01218bf8aaafa8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR