Triethyl orthoformate ≥98.0%

Supplier: TCI America
Danger

O0066-500ML O0066-25ML
TCO0066-500MLEA 61.17 USD
TCO0066-500ML TCO0066-25ML
Triethyl orthoformate ≥98.0%
Triethyl orthoformate

CAS Number: 122-51-0
MDL Number: MFCD00009230
Molecular Formula: C7H16O3
Molecular Weight: 148.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Melting point (°C): -76
Flash Point (°C): 35
Specific Gravity (20/20): 0.89


Product Categories: Synthetic Organic Chemistry, Formylation, Alkylation (Esterification)
Other Product Names / Synonyms: Ethyl Orthoformate, Orthoformic Acid Triethyl Ester, Triethoxymethane

Formula: C₇H₁₆O₃
MW: 148.2 g/mol
Boiling Pt: 145 °C
Melting Pt: –76 °C
Density: 0.89 (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009230
CAS Number: 122-51-0
EINECS: 204-550-4
UN: 2524
ADR: 3,III
Merck Index: 12,07012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac7722e46307866baa4a3c7268335d9e4d526551&vid=ac7722e46307866baa4a3c7268335d9e4d526551&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR