1-Octene ≥97.0%

Supplier: TCI America

O0041-25ML O0041-500ML
TCO0041-025MLEA 15.67 USD
TCO0041-025ML TCO0041-500ML
1-Octene ≥97.0%
1-Octene

CAS Number: 111-66-0
MDL Number: MFCD00009548
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 123
Melting point (°C): -101
Flash Point (°C): 10
Specific Gravity (20/20): 0.72
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Octylene

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123 °C
Melting Pt: –101 °C
Density: 0.72 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3,II
Merck Index: 13,01770
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d4d9d0da0fd320478a385772866e6267cc44f6d&vid=c1856fac6231093fe52d2f067bfb324ba8fd5559&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR