Suberic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid
O0023-25G
O0023-500G
O0023-100G
TCO0023-025GEA
25.43
USD
TCO0023-025G
TCO0023-500G
TCO0023-100G
Suberic acid ≥99.0% (by GC, titration analysis)
Suberic acid
CAS Number: 505-48-6
MDL Number: MFCD00004428
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 279
Melting point (°C): 143
Flash Point (°C): 210
Other Product Names / Synonyms: Cork Acid, 1,6-Hexanedicarboxylic Acid, Kork Acid, Octanedioic Acid
Formula:
C₈H₁₄O₄ MW: 174.2 g/mol Boiling Pt: 279 °C Melting Pt: 143 °C Density: 1.354 g/cm³ (21 °C) Flash Pt: 210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004428 CAS Number: 505-48-6 EINECS: 208-010-9 Merck Index: 12,09031 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...