Octadecylamine ≥85.0%

Supplier: TCI America
Danger

Synonyms: Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane

O0014-500G O0014-25G
TCO0014-500GEA 51.3 USD
TCO0014-500G TCO0014-25G
Octadecylamine ≥85.0%
Octadecylamine

CAS Number: 124-30-1
MDL Number: MFCD00008159
Molecular Formula: C18H39N
Molecular Weight: 269.52
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Boiling point (°C): 349
Melting point (°C): 53
Flash Point (°C): 148


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminooctadecane, Octadecylamine

Formula: H₃C(CH₂)₁₆CH₂NH₂
MW: 269.51 g/mol
Boiling Pt: 349 °C
Melting Pt: 53 °C
Density: 0.8618 g/cm³ (20 °C)
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00008159
CAS Number: 124-30-1
EINECS: 204-695-3
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e189ee24e52670f16ae2d3118eb55dad782377&vid=8392577880f0ccdbefef729ead64251a12a068bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR