Oleic acid ≥85.0%

Supplier: TCI America

Synonyms: (9Z)-9-Octadecenoic acid

O0011-500ML O0011-25ML
TCO0011-500MLEA 81.98 USD
TCO0011-500ML TCO0011-025ML
Oleic acid ≥85.0%
Oleic acid

CAS Number: 112-80-1
MDL Number: MFCD00064242
Molecular Formula: C18H34O2
Molecular Weight: 282.47
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 285
Flash Point (°C): 187
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: cis-9-Octadecenoic Acid

Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 285 °C
Melting Pt: 16.7 °C
Density: 0.89 (20 °C)
Flash Pt: 187 °C
Storage Temperature: Freezer
MDL Number: MFCD00064242
CAS Number: 112-80-1
EINECS: 204-007-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc11a024921ec32ee758b9ad9c2b2e9993756b5&vid=efc11a024921ec32ee758b9ad9c2b2e9993756b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR