1-Octadecene ≥90.0%

Supplier: TCI America

O0008-25ML O0008-500ML
TCO0008-025MLEA 19.76 USD
TCO0008-025ML TCO0008-500ML
1-Octadecene ≥90.0%
1-Octadecene

CAS Number: 112-88-9
MDL Number: MFCD00009003
Molecular Formula: C18H36
Molecular Weight: 252.49
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 149
Freezing point (°C): 16
Specific Gravity (20/20): 0.79

Formula: C₁₈H₃₆
Boiling Pt: 179 °C
Melting Pt: 17.5 °C
Density: 0.79 (20 °C)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS Number: 112-88-9
EINECS: 204-012-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a3a87e0ba6a21ea57192a0850b3ece30977fffc&vid=72dafb8b127f1ca3601d639b1a7574b0ccf91db5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR