4-Nitroindole ≥98.0%

Supplier: TCI America

Synonyms: 4-Nitro-1H-indole

N0628-5G N0628-1G
TCN0628-005GEA 106.7 USD
TCN0628-005G TCN0628-001G
4-Nitroindole ≥98.0%
4-Nitroindole

CAS Number: 4769-97-5
MDL Number: MFCD00010056
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 206

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00010056
CAS Number: 4769-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302cc716c26363b8b8538fb1f441c0bb036a3182&vid=302cc716c26363b8b8538fb1f441c0bb036a3182&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR