(3-Nitrophenyl)acetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
N0442-1G
N0442-5G
TCN0442-001GEA
24
USD
TCN0442-001G
TCN0442-005G
(3-Nitrophenyl)acetic acid ≥98.0% (by titrimetric analysis)
(3-Nitrophenyl)acetic acid
CAS Number: 1877-73-2
MDL Number: MFCD00007278
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 119
Formula:
(C₆H₄(NO₂))CH₂COOH Melting Pt: 119 °C |
MDL Number:
MFCD00007278 CAS Number: 1877-73-2 EINECS: 217-512-7 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...