6-Nitroindoline ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,3-Dihydro-6-nitro-(1H)-indole

N0403-5G N0403-1G
TCN0403-005GEA 75 USD
TCN0403-005G TCN0403-001G
6-Nitroindoline ≥97.0% (by GC, titration analysis)
6-Nitroindoline

CAS Number: 19727-83-4
MDL Number: MFCD00005710
Molecular Formula: C8H8N2O2
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 6-Nitro-2,3-dihydroindole

Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00005710
CAS Number: 19727-83-4
EINECS: 243-257-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284798489d2840f748e6c31773395d1ffca5e5b5&vid=284798489d2840f748e6c31773395d1ffca5e5b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR