4-Nitro-m-cresol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Methyl-4-nitrophenol, 1-Hydroxy-3-methyl-4-nitrobenzene
N0351-500G
N0351-25G
TCN0351-500GEA
364
USD
TCN0351-500G
TCN0351-25G
4-Nitro-m-cresol ≥98.0% (by GC, titration analysis)
4-Nitro-m-cresol
CAS Number: 2581-34-2
MDL Number: MFCD00007333
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 128
Other Product Names / Synonyms: 3-Methyl-4-nitrophenol
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007333 CAS Number: 2581-34-2 EINECS: 219-952-5 UN: 2446 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...