5-Nonanone ≥98.0%
Supplier: TCI America
Synonyms:
Nonan-5-one
N0294-500ML
N0294-25ML
N0294-100ML
TCN0294-500MLEA
543
USD
TCN0294-500ML
TCN0294-25ML
TCN0294-100ML
5-Nonanone ≥98.0%
5-Nonanone
CAS Number: 502-56-7
MDL Number: MFCD00009484
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 60
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: Dibutyl Ketone, Valerone
Formula:
C₉H₁₈O Boiling Pt: 186 °C Melting Pt: -50 °C Density: 0.82 (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009484 CAS Number: 502-56-7 EINECS: 207-946-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...