1-Nonanol ≥99.0%

Supplier: TCI America
Warning

Synonyms: Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol

N0292-25ML N0292-500ML
TCN0292-025MLEA 20 USD
TCN0292-025ML TCN0292-500ML
1-Nonanol ≥99.0%
1-Nonanol

CAS Number: 143-08-8
MDL Number: MFCD00002990
Molecular Formula: C9H20O
Molecular Weight: 144.26
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 104
Freezing point (°C): -5
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Nonyl Alcohol

Formula: H₃C(CH₂)₇CH₂OH
MW: 144.26 g/mol
Boiling Pt: 214 °C
Melting Pt: –7 °C
Density: 0.83 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002990
CAS Number: 143-08-8
EINECS: 205-583-7
UN: 3082
ADR: 9,III
Merck Index: 12,06774
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e176a40da40d6563dac18750605e7a2d8d4ab06f&vid=7380dcc2ea3eea7a13c5184ffd94391be4fc2c99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR